Learn More
ACROBiosystems Flt-3 Ligand 100ug
Supplier:  ACROBiosystems FLDR52H3100UG
Encompass_Preferred
Rhinopithecus bieti Flt-3 Ligand Protein His Tag (FLD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 27 -Pro 184 (Accession # A0A2K6LTJ0).
Catalog No. 50-253-5060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.