Learn More
MDI Membrane FLT MDI PTFE+GF 25MM .45 100PK
Supplier:  MDI Membrane SYTG0602MNXX104
Encompass_Preferred
mdi PTFE Syringe Filter with multi-layered Glass Fiber prefilter, 25 mm, 0.45 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8737
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.