Learn More
Cytiva (Formerly Pall Lab) Versaflow Capsule 0.8/0.45 m, Sterile (1/pkg)
Supplier:  Cytiva 12131
Encompass_Preferred
Membrane capsule is designed to filter liquids not requiring sterilization, or as a prefilter before final sterilization-For use before final sterilization-Porous membrane allows high flow rate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3187
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.