Learn More
Parker Hannifan FLTR CPSL PREPOR PES 3PK
SDP
Supplier:  Parker Hannifan FEPSA020GGPS3
Encompass
Parker domnick hunter Demicap Filter Capsules, Prepor PES Membrane - 0.2um Pore Size, Filtration Area: 0.25m2, Inlet Style: Stepped Hosebarb, Length: 200mm, 7.87 in., Sterile, Material: Polyethersulfone, Pore Size: 0.2um, Autoclavable: Yes
Catalog No. NC1917822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.