Learn More
Parker Hannifan FLTR CPSL PROPOR PES 3PK
SDP
Supplier:  Parker Hannifan FEMSB020GGPSX
Encompass_Preferred
Parker domnick hunter Demicap Filter Capsules, Propor PES Membrane - 0.2micrometer Pore Size, Polypropylene, Filtration Area: 0.11m2, Length: 5.5 in., 140mm, Outlet Style: Stepped Hosebarb, Material: Polyethersulfone; Polypropylene, Pore Size: 0.2um, Autoclavable: Yes
Catalog No. NC1917815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.