Learn More
Cytiva (Formerly Pall Lab) 47 mm Pressure Filtration Funnel, Stainless Steel - 47 mm pressure filtration funnel, stainless steel (1/pkg)
Supplier:  Cytiva 4280
Encompass_Preferred
High Quality, Corrosion-Resistant Funnel for Rapid Batch Filtration-Stainless steel construction offers excellent chemical resistance. -Convenient hand tightening makes filter changing easy. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3025
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.