Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluconazole, 99%

Catalog No. AAJ6201506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6201506 5 g
AAJ6201503 1 g
AAJ6201514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6201506 Supplier Thermo Scientific Chemicals Supplier No. J6201506
Add to Cart

Fluconazole, CAS # 86386-73-4, is a synthetic triazole with antifungal activity against some species of Candida. It also shows inhibitory activity against cytochrome P-450 enzymes and it is a demethylase. | CAS: 86386-73-4 | C13H12F2N6O | 306.277 g/mol

Fluconazole is used to treat a variety of fungal infections, especially Candida infections of the vagina (yeast infections), mouth, throat and bloodstream. It is also used by the people having weak immune system such as persons with neutropenia due to cancer chemotherapy, transplanted patients and premature infants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Fluconazole, 98% is a triazole compound showing antifungal activity mainly against Candida, Coccidioides, and Cryptococcus species
  • It has been shown to inhibit fungal cytochrome P-450 sterol C-14 α-demethylation, resulting in the accumulation of fungal 14 α-methyl sterols, the loss of normal fungal sterols, and associated fungistatic activity
  • It also inhibits cytochrome P450 family 2 subfamily c member 9 (CYP2C9)
  • Mammalian cell demethylation is much less susceptible to fluconazole inhibition

Applications

  • Fluconazole has fungistatic activity against many fungal strains including Candida albicans, Candida glabrata, Candida parapsilosis, Candida tropicalis, and Cryptococcus neoformans.
  • Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene
  • In biological experiments, this compound has been used to assess minimum inhibitory concentration in (MIC)-fungal assays
  • It has also been used as a positive control in antifungal assay of plants
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86386-73-4
Molecular Formula C13H12F2N6O
Molecular Weight (g/mol) 306.277
MDL Number MFCD00274549
InChI Key RFHAOTPXVQNOHP-UHFFFAOYSA-N
Synonym fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol
PubChem CID 3365
ChEBI CHEBI:46081
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
SMILES C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O

Specifications

Melting Point 138°C to 140°C
Color White
Density 1.05
Quantity 5 g
Merck Index 14,4122
Solubility Information Soluble indimethylsulfoxide and ethanol.
Formula Weight 306.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Fluconazole
Hazard Category H302-H315-H319-H360D
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
RTECSNumber XZ4810000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.