Learn More
Abcam Fluconazole, Cytochrome P450 Inhibitor, >99% Purity, Solid, 50 mg
Description
- Purity: >99%
- Molecular Weight: 306.27 Da
- Biological Activity: Triazole antifungal agent; Cytochrome P450 inhibitor (IC50 values are 30.3, 12.3, and 13.1 μM for CYP2C9, CYP2C19, and CYP3A4, respectively)
- Key Characteristic: Penetrates the blood-brain barrier
- Physical Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Cytochrome P450 (CYP2C9, CYP2C19, CYP3A4) |
| Buffer | 100% Fluconazole |
| Formulation | 100% Fluconazole |
| Quantity | 50 mg |
| For Use With (Application) | Cytochrome P450 inhibition, Antifungal research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.