Learn More
Fluotics Tecan Fluent COMBI Compatible P1000, sterile
Supplier: Fluotics CLHA10CBR.F
Tecan Fluent COMBI Compatible P10 filter/sterile Working Volume 10uL Tip Color Black Single Stack COMBI MCA LiHa/FCA 24 racks/case with 96 tips per rack