Learn More
Fluotics Tecan Fluent COMBI Compatible P50, filter/sterile
Supplier:  Fluotics CLHA50R.ST
Encompass
Tecan Fluent COMBI Compatible P50 sterile Working Volume 50uL Tip Color Clear Single Stack COMBI MCA LiHa/FCA 24 racks/case with 96 tips per rack
Catalog No. 50-004-35265