Learn More
Apexbio Technology LLC Flufenamic acid 530-78-9 50mg
SDP
Supplier:  Apexbio Technology LLC B176050
Encompass_Preferred
Flufenamic acid (CAS 530-78-9) is a small-molecule inhibitor targeting cyclooxygenase (COX) enzymes and TEAD transcription factors It is designed to inhibit COX-mediated prostaglandin synthesis and block TEAD-YAP-dependent signaling thereby regulating inflammatory responses and gene transcription Flufenamic acid exerts its biological activity primarily through inhibition of COX enzymes and blockade of ion channels including chloride channels (IC50 10 20 M) and L-type calcium channels (IC50 10 M) as well as activation of specific potassium channels The compound also binds the YBD domain of TEAD2 inhibiting YAP-TEAD pathway-dependent cellular proliferation and migration Based on these pharmacological properties flufenamic acid holds research potential in studies of inflammatory pathways ion channel physiology electrophysiological responses and TEAD-mediated signaling
Catalog No. 50-224-0831
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.