Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flufenamic acid, MilliporeSigma™ Supelco™

Catalog No. 111019379 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9379 50 g
11-102-0607 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9379 Supplier MilliporeSigma Supelco Supplier No. F900550G
Add to Cart
Add to Cart

Flufenamic acid, a non-steroidal anti-inflammatory drug, is an effective analgesic agent.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 530-78-9
Physical Form Neat
Linear Formula 2-(CF3C6H4 NH)C6H4 CO2 H
MDL Number MFCD00002422
Melting Point 132°C to 135°C
Synonym N-(3-[Trifluoromethyl]phenyl)anthranilic acid; N-(alpha,alpha,alpha-Trifluoro-m-tolyl)anthranilic acid
Type Analytical Standard
Industry Type Drug Analysis
Quantity 50 g
Formula Weight 281.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.