Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Description
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
Fluid Thioglycollate Medium is available as dehydrated culture medium or in ready-to-use prepared media tubes and bottles and in formats that meet international industrial guidelines for microbiological testing, including United States Pharmacopeia (USP) guidelines for sterility testing.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, COMPF, USP |
| Color | Clear broth w/visible agar light to medium amber |
| Content And Storage | 2 to 25°C |
| Format | Bottle |
| Manufacturing Origin | United States |
| Media Type | Fluid Thioglycollate Medium |
| Product Type | Microbiology Media |
| Fill Volume | 100 mL |
| Includes | w/ Indicator, Dextrose Medium |
| Packaging Type | Glass Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.