Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Description
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
| Dimensions | 1/8 in. ID x 1/4 in. OD to 3/8 in. ID x 5/8 in. OD |
| Tubing Style | 3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 2 m (6.6 ft); Luer to MPC |
| Connector Types | Luer to MPC |
| Product Type | Adapter Set |
| Tubing Size | TPE 374, 2 m (6.6 ft) |