Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Description
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
| Tubing Style | 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
| Connector Types | MPX to MPC |
| Product Type | Adapter Set |
| Tubing Size | TPE 374, 2 m (6.6 ft) |