Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
GREENER_CHOICE
Catalog No. SH3111201
Change view
Click to view available options
Tubing Size:
10 in. TPE 374, 19.7 in. Pharmed BPT
Connector Types:
Luer
MPC
Dimensions:
1/4 in. ID x 3/8 in. OD
1/4 in. ID x 7/16 in. OD
1/8 in. ID x 1/4 in. OD
3/8 in. ID x 5/8 in. OD
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tubing Size Connector Types Dimensions
SH3111201 10 in. TPE 374, 19.7 in. Pharmed BPT MPC 3/8 in. ID x 5/8 in. OD
SH3111101 10 in. TPE 374, 19.7 in. Pharmed BPT Luer 1/8 in. ID x 1/4 in. OD
SH3111501 10 in. TPE 374, 19.7 in. Pharmed BPT MPC 1/4 in. ID x 7/16 in. OD
SH3111401 10 in. TPE 374, 19.7 in. Pharmed BPT MPC 1/4 in. ID x 3/8 in. OD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3111201 Supplier Thermo Scientific™ Supplier No. SH31112.01
Add to Cart
Add to Cart
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 3/8 in. ID x 5/8 in. OD
Fluid Geometry at Working Volume 3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m(10 in.); TPE 374; Luer
Tubing Style 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25m (10 in.); TPE 374; MPC
Connector Types MPC
Product Type Pump Set
Tubing Size 10 in. TPE 374, 19.7 in. Pharmed BPT