Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Description
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
| Dimensions | 1/4 in. ID x 3/8 in. OD |
| Tubing Style | 6.35 mm (1/4 in.) ID x 9.52 mm (3/8 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m (10 in.); TPE 374; MPC |
| Connector Types | MPC |
| Product Type | Pump Set |
| Tubing Size | 10 in. TPE 374, 19.7 in. Pharmed BPT |