Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Description
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
| Dimensions | 1/2 in. ID x 3/4 in. wall |
| Tubing Style | 3 x TPE 374 |
| Connector Types | MPX body-insert |
| Product Type | Y Set |
| Tubing Size | 2 x 20 in. to 20 in. (Length) |