Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FluidEase™ Pro ClipTip™ Electronic Multichannel Pipettes
GREENER_CHOICE

Catalog No. 01671135
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 30 μL
10 to 300 μL
2.5 to 100 μL
25 to 1250 μL
3 to 125 μL
5 to 200 μL
Number of Channels:
8
12
16
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
01-671-135 2.5 to 100 μL 12
01-671-138 0.5 to 10 μL 8
01-671-136 2.5 to 100 μL 8
01-671-134 5 to 200 μL 8
01-671-132 10 to 300 μL 8
01-671-130 25 to 1250 μL 8
01-671-137 0.5 to 10 μL 12
01-671-133 5 to 200 μL 12
01-671-131 10 to 300 μL 12
01-671-129 25 to 1250 μL 12
01-671-128 0.5 to 10 μL 16
01-671-127 1 to 30 μL 16
01-671-126 3 to 125 μL 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 01-671-135 Supplier Thermo Scientific™ Supplier No. 4676040
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes enable effortless 96- and 384-well plate filling with touchscreen control, ultra-wide speed selection, and secure ClipTip™ sealing for reproducible, high-throughput workflows in shared labs.

Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes are designed to simplify and standardize plate-based workflows. A large touchscreen interface and ultra-wide speed selection support precise aspiration and dispense across liquid types. Secure ClipTip™ sealing, effortless column-to-row transitions, preset and advanced programming modes, and password-protected settings enable reproducible, high-throughput plate filling across shared laboratory environments

Features:

  • Large Touchscreen User Interface: A high-resolution touchscreen provides direct, visual interaction for setup and execution, reducing training time and minimizing setup errors compared to button- or joystick-based navigation.
  • Ultra-Wide Speed Selection: One of the broadest aspiration and dispense speed ranges available supports gentle handling of sensitive samples, precise control of viscous liquids, and faster execution for time-critical workflows.
  • Simple Presets and Advanced Programming: Easy-to-use preset modes support fast onboarding for new or occasional users, while advanced step programming enables complex, multi-step protocols for demanding applications.
  • Index-Finger Operation with Increased Rotational Flexibility: Retains Thermo Fisher–recognized index-finger operation with a wider angle of rotation at the finger rest to accommodate different hand sizes, grip angles, and working positions.
  • Secure ClipTip™ Technology: ClipTip™ tips lock and seal in place until intentional ejection, helping ensure consistent volume delivery and reduced variability from dispense to dispense.
  • Electronic Tip Ejection with Safe-to-Eject Illumination: Electronic tip ejection reduces physical effort and includes LED illumination to indicate when ejection is possible, providing ergonomic benefit and added user security.
  • Password Protection and User Profiles: Password-protect critical settings and protocols and support up to five user profiles to enable standardized operation in shared and regulated laboratory environments.
  • Custom Calibration Options: Enable calibration to customer-specific liquids or environmental conditions to align performance with real laboratory workflows.
  • Service and Maintenance Tracking: Built-in service reminders, calibration logs, and usage tracking support instrument management and compliance documentation over time.
TRUSTED_SUSTAINABILITY

Specifications

Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Yellow
Disposable Reusable
For Use With ClipTip 200
Model Bluetooth
Number of Channels 12
Programmability Yes
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 2.5 to 100 μL
Display Touchscreen
Finger Style Ergonomic Index Finger
Multilingual User Interface Yes
Warranty 1 Year, 2 Years with Registration
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.