Learn More
Sartorius iQue Fluidics Maintenance Kit Screener Plus, Prevents Downtime, Maintains Accurate Results, 1 kit
Catalog No. NC1246825 Shop All Sartorius Products
Encompass
The iQue Fluidics Maintenance Kit Screener Plus is an all-in-one solution for routine maintenance of Sartorius flow cytometry systems, ensuring optimal performance and longevity. Regularly using this kit helps maintain data quality and prevents costly downtime. It's designed for hassle-free maintenance with essential components tailored for iQue 3 and iQue Screener Plus, supporting bi-monthly servicing or maintenance after 176 hours of use.
Catalog No. NC1246825 Supplier Sartorius Supplier No. 91094
The iQue Fluidics Maintenance Kit Screener Plus is an all-in-one solution for routine maintenance of Sartorius flow cytometry systems, ensuring optimal performance and longevity. Regularly using this kit helps maintain data quality and prevents costly downtime. It's designed for hassle-free maintenance with essential components tailored for iQue 3 and iQue Screener Plus, supporting bi-monthly servicing or maintenance after 176 hours of use.