Learn More
Honeywell™ Fluka™ Inert II SPE, Empty Columns
Catalog No. 6002889 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Column Capacity (Metric):
8 mL
25 mL
Description:
Bulk
Loose frits
No Frits
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Capacity (Metric) Description
600-28-89 8 mL Bulk
600-30-41 25 mL Loose frits
600-30-42 8 mL No Frits
600-30-43 8 mL Loose frits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 600-28-89 Supplier Honeywell™ Supplier No. 5435
Inert plastic housing with PTFE frits
  • Reduced background contamination results in significant gains in detection capabilities
  • Decreases downtime for GC/MS source cleaning
  • Eliminates extraneous peaks
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 8 mL
Description Bulk
Type Empty Solid Phase Extraction Column
Quantity 1000