Description
- Reduced background contamination results in significant gains in detection capabilities
- Decreases downtime for GC/MS source cleaning
- Eliminates extraneous peaks
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
Specifications
Specifications
| Column Capacity (Metric) | 25 mL |
| Description | Loose frits |
| Type | Empty Solid Phase Extraction Column |
| Quantity | 300 |