Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Fluka™ Non-Polar Silica Based SPE Columns
End-capped, Medical-grade polypropylene with PE frits
Supplier: Honeywell™ 9034
Description
- All columns are reversed phase and should be preconditioned with a polar solvent with the elution solvent being non-polar
- All columns are endcapped and have a surface area of 500m2/g

Specifications
6 mL | |
40 to 63 μm | |
Yes | |
30 |
500 mg | |
Ethyl (C2) | |
60 Å | |
500 m2/g |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction