Learn More
Honeywell™ Fluka™ QuEChERS, Sample Cleanup
Centrifuge Tube for Dispersive SPE
Supplier: Honeywell™ 5651
Description
- Efficient method of sample preparation
- Array of tube sizes and sorbent materials available
- Appropriate for use with Burdick and Jackson acetonitrile and other high purity solvents
Specifications
| Dispersive Solid Phase Extraction | |
| 150 mg MgSO4, 50 mg PSA | |
| 2 mL | |
| 100 |