Description
- Efficient method of sample preparation
- Array of tube sizes and sorbent materials available
- Appropriate for use with acetonitrile and other high purity solvents
Specifications
Specifications
| For Use With (Application) | Dispersive Solid Phase Extraction |
| Sorbent | 150 mg MgSO4, 150 mg PSA, 50 mg C18 |
| Volume (Metric) | 2 mL |
| Quantity | 100 |