Learn More
Honeywell™ Fluka™ QuEChERS, Sample Cleanup
Centrifuge Tube for Dispersive SPE
Supplier:  Honeywell™ 5658
 View more versions of this product
Catalog No. 600-29-58
Description
Description
  • Efficient method of sample preparation
  • Array of tube sizes and sorbent materials available
  • Appropriate for use with Burdick and Jackson acetonitrile and other high purity solvents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dispersive Solid Phase Extraction
150 mg MgSO4, 25 mg PSA, 7.5 mg GCB
2 mL
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.