Learn More
Honeywell™ Fluka™ QuEChERS, Sample Extraction
Extraction Salt for QuEChERS Method with 50 mL Tube
Supplier:  Honeywell™ 5602
 View more versions of this product
Catalog No. 600-29-69
Description
Description
QuEChERS
  • Quick, Easy, Cheap, Effective, Rugged and Safe Sample Extraction and Cleanup Products
  • Honeywell Burdick & Jackson™ QuEChERS products offer an alternative sample preparation technique to traditional liquid-liquid and solid phase extraction (SPE) methods
  • the use of QuEChERS has the potential to lower the overall cost of sample preparation due to the increased efficiency and ease-of use when compared with SPE methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN15662 QuEChERS Method
4 gm MgSO4, 1 gm sodium chloride, 500 mg sodium citrate (dibasic sesquihydrate),1 gm sodium citrate (tribasic dihydrate) per Tube
50 mL
50
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.