Learn More
Cole-Parmer Fluke Calibration 7103 Portable Calibration Bath; -30 to 125C
Supplier:  Cole-Parmer 9091204
Encompass
These calibration baths are designed with the stability and accuracy of a fluid bath and the compact size and weight of a portable calibrator. Smaller and lighter than most dry-well calibrators, these calibration baths are easy to carry and are spill-proof. The heating, cooling, and stabilization times allow you to take readings within 45 minutes. Bath is ideal for glass and bimetal thermometers. There’s no need to make custom well inserts — the 2-½" diameter stirred tank holds just about any size probe. All calibration baths have built-in RS-232 interface and include software for Windows 3.x/95. The basic data collection software allows you to change the temperature, set points, and safety cutouts on your bath. Available for sale in the United States and Canada only.
Catalog No. NC2175435
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.