Learn More
AdipoGen Flumetsulam
SDP
Supplier:  AdipoGen CDXF0080G001
Encompass_Preferred
Chemical. CAS 98967-40-9. Formula C12H9F2N5O2S. MW 325.3. Synthetic. Highly active herbicide of the triazolopyrimidine sulfonanilide class. Flumetsulam acts by inhibiting acetolactate synthase. Flumetsulam exhibits broad spectrum activity on broadleaf weed species with selectivity to several major agronomic crops in soil and foliar applications. Compound can be used as analytical reference material.
Catalog No. 50-256-0893
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.