Learn More
AdipoGen Flumiclorac
SDP
Supplier:  AdipoGen CDXF0085G001
Encompass_Preferred
Chemical. CAS 87547-04-4. Formula C16H13ClFNO5. MW 353.73. Synthetic Herbicide. Mode of action is similar to herbicides that inhibit the protoporphyrinogen oxidase Protox enzyme in susceptible plant species that leads to an uncontrolled nonenzymatic oxidation of protoporphyrinogen IX Protogen to protoporphyrin IX Proto and successive necrosis.
Catalog No. 50-256-0885
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.