Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fluo-2 AM, Cell Permeable, Calcium Binding Dye, 1 mg

Catalog No. 502706186
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6186 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6186 Supplier Abcam Supplier No. AB1427751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High affinity, cell-permeable green fluorescent dye for intracellular Ca2+ binding.

  • High affinity, green fluorescent Ca²± binding dye
  • Cell-permeable
  • Binds to intracellular Ca²± with Kd = 230 nM
  • Fluorescence intensity increases upon Ca²± binding
  • Excitation: 488 nm; Emission: 515 nm
  • Molecular Weight: 1061 Da
  • Purity: >95%

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Calcium (Ca2+)
Substrate Properties Chemical Substrate
Substrate Ca2+
Shipping Condition Ambient - Can Ship with Ice
Content And Storage Store at -20°C.
Detection Location In-Cell Detection
Form Solid
Detection Method Fluorescence
Formulation 95% Fluo-2, AM
Quantity 1 mg
For Use With (Application) Calcium imaging, Cell-based assays
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.