Learn More
Tocris Bioscience™ Fluo-4 AM



Description
Specifications
Specifications
| Chemical Name or Material | N-[4-[6-[(Acetyloxy)methoxy]-2,7-difluoro-3-oxo-3H-xanthen-9-yl]-2-[2-[2-[bis[2-[(acetyloxy)methoxy]-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-N-[2-[(acetyloxy)methoxy]-2-oxoethyl]glycine (acetyloxy)methyl ester |
| CAS | 273221-67-3 |
| Quantity | 1 mg |
| Target | Calcium Signaling Probes and Ca2+ Indicator Dyes |
| Molecular Formula | C51H50F2N2O23 |
| Formula Weight | 1096.95 |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.