Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fluo 4 Assay Kit, Calcium, Fluorescent, 10 Unit

Catalog No. 502587022
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-7022 10 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-7022 Supplier Abcam Supplier No. AB22855510UNIT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent assay kit for measuring calcium in suspension and adherent cell samples.

  • Detection method: Fluorescent
  • Sample types: Suspension cells, Adherent cells
  • Cited in: 5 publications
  • Usage: Suitable for use with Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Calcium
Substrate Properties Chemical Substrate
Substrate Fluo-4
Label or Dye Fluo-4
For Use With (Equipment) Microplate Reader
Format Kit
Detection Location In-Cell Detection
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 10 Units
For Use With (Application) Calcium measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.