Learn More
Abcam Fluo-8 Calcium Assay Kit, Cell-based, Fluorescent, 10x96 Tests
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Sample types: Suspension cells, Adherent cells
- Instrumentation: Microplate reader
- Applications: Calcium measurement
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | Fluo-8 |
| Detection Method | Fluorescence |
| Detection Location | In-Cell Detection |
| Target Molecule | Calcium |
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 960 Tests |
| Substrate Properties | Chemical Substrate |
| Format | 96-well Plate Kit |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 10 x 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.