Learn More
Abcam Fluo 8 Calcium Assay Kit, Medium Removal, 100x96 Tests
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Sample types: Suspension cells, Adherent cells
- Equipment compatibility: Microplate reader
- Research citations: Cited in > 2 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Calcium |
| Substrate Properties | Chemical Substrate |
| Format | Kit |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Microplate Reader |
| Label or Dye | Fluo-8 |
| No. of Reactions | 9600 Tests |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 100 x 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.