Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Fluo-8 Calcium Assay Kit, Medium Removal, Cell-based, 1x96 Tests
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Sample types: Suspension cells, Adherent cells
- Use: Microplate reader
Specifications
Specifications
| Target Molecule | Calcium |
| Substrate Properties | Chemical Substrate |
| No. of Reactions | 96 Tests |
| Label or Dye | Fluo-8 |
| For Use With (Equipment) | Microplate Reader |
| Detection Location | In-Cell Detection |
| Format | Kit |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 96 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?