Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinolone Acetonide, Anhydrous, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18602767 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
250 g
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-767 100 g Amber Glass Bottle
18-602-764 250 mg Amber Glass Bottle
18-602-765 1 g Amber Glass Bottle
18-602-766 5 g Amber Glass Bottle
18-602-769 25 g Amber Glass Bottle
18-602-768 250 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-767 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1118100GMBL
Add to Cart
Add to Cart
Request a Quote

F1118, 67-73-2, C24H30F2O6

Spectrum™ Chemical Fluocinolone Acetonide, Anhydrous, Micronized, USP is a corticosteroid primarily used in dermatology to reduce skin inflammation and relieve itching. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-73-2
Molecular Formula C24H30F2O6
Molecular Weight (g/mol) 452.50
MDL Number MFCD00010525
InChI Key FEBLZLNTKCEFIT-VSXGLTOVSA-N
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

CAS Max % 100%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +98° to +108°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fluocinolone Acetonide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.