Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinolone acetonide, Thermo Scientific Chemicals

Catalog No. AAJ6399603
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6399603 1 g
AAJ63996MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6399603 Supplier Thermo Scientific Chemicals Supplier No. J6399603
Add to Cart

A glucocorticoid with anti-inflammatory properties

Fluocinolone acetonide is primarily used in dermatology to reduce skin inflammation and relieve itching. It acts as an inhibitor of tumor cells and it can regulate lipid metabolism by modulating gene expression. It can also inhibit the expression of VEGF (vascular endothelial growth factor) in the retinal pigment epithelial cell line due to its high glucocorticoid receptor affinity. Furthermore, it can inhibit TNF-α angiogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluocinolone acetonide is primarily used in dermatology to reduce skin inflammation and relieve itching. It acts as an inhibitor of tumor cells and it can regulate lipid metabolism by modulating gene expression. It can also inhibit the expression of VEGF (vascular endothelial growth factor) in the retinal pigment epithelial cell line due to its high glucocorticoid receptor affinity. Furthermore, it can inhibit TNF-α angiogenesis.

Solubility
Soluble in water (partly), DMSO, alcohol, chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-73-2
Molecular Formula C24H30F2O6
Molecular Weight (g/mol) 452.50
MDL Number MFCD00010525
InChI Key FEBLZLNTKCEFIT-VSXGLTOVSA-N
Synonym fluocinolone acetonide, synalar, flucinar, synandone, fluonid, jellin, synamol, synemol, coriphate, fluovitif
PubChem CID 6215
ChEBI CHEBI:31623
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Melting Point 267°C to 269°C
Color White
Quantity 1 g
Merck Index 14,4150
Solubility Information Soluble in water (partly),DMSO,alcohol,chloroform and methanol.
Formula Weight 452.49
Physical Form Crystalline Powder
Chemical Name or Material Fluocinolone acetonide
Hazard Category H315-H319-H335-H361
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 200-668-5
RTECSNumber TU3830000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.