Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinonide, Micronized, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18602770 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
250 g
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-770 250 mg Amber Glass Bottle
18-602-771 1 g Amber Glass Bottle
18-602-772 5 g Amber Glass Bottle
18-602-773 25 g Amber Glass Bottle
18-602-775 100 g Amber Glass Bottle
18-602-774 250 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-770 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1123250MG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

F1123, 356-12-7, C26H32F2O7

Spectrum™ Chemical Fluocinonide, Micronized, USP is glucocorticoid steroid used as an anti-inflammatory treatment for disorders like seborrheic dermatitis or eczema. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 356-12-7
Molecular Formula C26H32F2O7
Molecular Weight (g/mol) 494.53
InChI Key WJOHZNCJWYWUJD-IUGZLZTKSA-N
IUPAC Name 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C

Specifications

CAS Max % 100%
Melting Point 308 to 311°C
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 250 mg
UN Number UN2811
Specific Rotation +81° to +89°
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fluocinonide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.