Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorene-2-boronic acid pinacol ester, 95%

Catalog No. p-7034277
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5194703 1 g
AAH5194706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5194703 Supplier Thermo Scientific Chemicals Supplier No. H5194703
Only null left
Add to Cart

CAS: 922706-40-9 | C19H21BO2 | 292.185 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922706-40-9
Molecular Formula C19H21BO2
Molecular Weight (g/mol) 292.185
MDL Number MFCD16294548
InChI Key WVJQQLZHOADEAF-UHFFFAOYSA-N
Synonym 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-2-fluorenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fluorene-2-boronic acid pinacol ester, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl, amtb249, 4,4,5,5-tetramethyl-2-9h-fluorene-2-yl-1,3,2-dioxaborolane, 2-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 56925000
IUPAC Name 2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3

Specifications

Quantity 1 g
Formula Weight 292.18
Percent Purity 95%
Chemical Name or Material Fluorene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.