Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorene-d10, SPEX CertiPrep™
SDP

Catalog No. S2036 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Amber Prescored Glass Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S-2036 1 mL Amber Prescored Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S-2036 Supplier SPEX CertiPrep Supplier No. S2036

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Fluorene-d1081103-79-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 81103-79-9 , 67-56-1
Molecular Formula C13H10
Molecular Weight (g/mol) 176.284
InChI Key NIHNNTQXNPWCJQ-QNNBDYQESA-N
Synonym fluorene-d10, 2 h?? fluorene, fluorene d10, fluorene-d10, analytical standard, fluorene d10 10 microg/ml in cyclohexane, fluorene-d10, 98 atom % d cp
PubChem CID 6913203
IUPAC Name 1,2,3,4,5,6,7,8,9,9-decadeuteriofluorene
SMILES C1C2=CC=CC=C2C3=CC=CC=C31

Specifications

Quantity 1 mL
Packaging Amber Prescored Glass Ampule
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Chemical Name or Material Fluorene-d10
Hazard Category Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.