Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fluorescein, Disodium Salt
SCI_ED

Catalog No. S25328A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25328A 100 g Powder
S25328 25 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25328A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stain and Indicator

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
Synonym Uranin, Disodium Salt
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Assay Percent Range 100% w/v
Linear Formula C20H10O5Na2
Formula Weight 412.3
Packaging Powder
Color Orange/Rust
Quantity 100 g
Chemical Name or Material Fluorescein, Disodium Salt
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.