CAS RN 518-47-8
Uranine (Concentrated, Water Soluble/Laboratory), Fisher Chemical™
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Fluorescein Sodium Salt IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Thermo Scientific Chemicals Fluorescein disodium salt hydrate
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Acid Yellow 73; C.I. 45350 IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fluorescein disodium salt
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Acid Yellow 73; C.I. 45350 IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fluorescein Sodium Salt, Sigma-Aldrich
CAS: 518-47-8 Molecular Weight (g/mol): 376.27 g/mol
Thermo Scientific Chemicals Fluorescein, disodium salt, pure
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fluorescein Sodium, Spectrum™ Chemical
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Uranine, TCI America™
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Fluorescein Sodium Salt IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagents
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Disodium Fluorescein, Uranine, Color Index 45350 IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Solstice
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Fisher Science Education™ Fluorescein, Disodium Salt
CAS: 518-47-8 Molecular Formula: C20H10Na2O5 Molecular Weight (g/mol): 376.28 MDL Number: MFCD00167039 InChI Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L Synonym: Uranin,Disodium Salt IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12