Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody
GREENER_CHOICE

Catalog No. A889
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A889 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A889 Supplier Invitrogen™ Supplier No. A889
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Fluorescein/Oregon Green Polyclonal Antibody for Flow, ICC/IF, IHC

Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein/Oregon Green
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M potassium phosphate with 5mM sodium azide; pH 8
Gene Alias FITC
Host Species Rabbit
Immunogen Fluorescein/Oregon Green.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?

One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.