Learn More
Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488

Description
The Alexa Fluor 488 conjugates can be used to amplify the fluorescence signal while still allowing fluorescein-compatible optics. Excitation 495 nm/peak emission 519 nm. It is a good practice to centrifuge the labeled antibody solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
Specifications
Specifications
| Antigen | Fluorescein/Oregon Green |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | 0.1M potassium phosphate with 5mM sodium azide; pH 8 |
| Gene Alias | FITC |
| Host Species | Rabbit |
| Immunogen | Fluorescein/Oregon Green. |
| Purification Method | Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.