Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. A11096
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11096 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11096 Supplier Invitrogen™ Supplier No. A11096
Add to Cart
Edge
Add to Cart

Goat Polyclonal Antibody

We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes. In addition to their utility in quenching the fluorescence of extracellular fluorescein, the high affinity of Invitrogen's anti-fluorescein antibodies for both fluorescein and the structurally related Oregon GreenTM dye makes them ideal for immunochemical applications. Harmer and Samuel report the fluorescein-anti-fluorescein system provides a sensitive alternative to the biotin-streptavidin methods conventionally used for enzyme-linked immunosorbent assays (ELISAs). Fluorescein has also been found to be an excellent hapten for FISH, yielding sensitive results with extremely low background levels. The Alexa Fluor 488 conjugate can be used to amplify the fluorescence signal while still allowing fluorescein-compatible optics. The approximate absorption (Abs) and fluorescence emission (Em) maxima in nm: 495/519. Our anti-dye antibodies can be used in many different applications. It is a good practice to centrifuge the labeled antibody solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any...

Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein/Oregon Green
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor 488
Formulation 0.1M potassium phosphate with 5mM sodium azide; pH 8
Gene Alias FITC
Host Species Goat
Immunogen Fluorescein/Oregon Green.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.