Learn More
Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody, HRP

Description
Storage and reconstitution: The formulation of this antibody was updated from PBS with 0.02% thimerosal to PBS without thimerosal on 31/Mar/2024, any lots produced after this date will not contain thimerosal. Lot numbers greater than 2855945 will not have thimerosal. Store product at -20°C until opened. Reconstitute using 0.5 mL PBS, pH 7.2, to yield a 1 mg/mL stock solution. After reconstitution in buffer without a stabilizer like thimerosal, the product is stable for approximately three months at 4°C. For prolonged storage after reconstitution in buffers without a stabilizer, add glycerol to a final concentration of 50% (v/v), aliquot, and store at -20°C. The antibody will be stable for up to one year stored in this manner. Alternatively, if the product is reconstituted in a buffer containing a preservative, for example, in PBS, pH 7.2, containing 0.02% thimerosal, the product will be stable for up to one year when stored at 4°C.
Specifications
Specifications
| Antigen | Fluorescein/Oregon Green |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Multiplex Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | 0.1M potassium phosphate with no preservative; pH 8 |
| Gene Alias | FITC |
| Host Species | Rabbit |
| Immunogen | Fluorescein/Oregon Green. |
| Purification Method | IgG fraction |
| Quantity | 500 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.