Learn More
Fluorescein-Ubiquitin
Description
For use in combination with FRET donors including LanthaScreen™ Tb-Ubiquitin (PV4375) or a LanthaScreen™ antibody including Tb-anti-GST (PV4216) or Tb-anti-ubiquitin-FK2 (PV4751).
Specifications
Specifications
| Quantity | 500 μg |
| Conjugate | Fluorescein |
| Content And Storage | Fluorescein-labeled ubiquitin in HEPES buffered saline. Store at -20°C. |
| Detection Method | Fluorescence |
| Excitation/Emission | 340⁄490, 520 |
| For Use With (Equipment) | Microplate Reader |
| Label or Dye | FITC (Fluorescein) |
| Label Type | Classic Dyes |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.