Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoresceinamine Isomer II 97%, Thermo Scientific™

Catalog No. AC191650010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC191650010 1 g
AC191650050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191650010 Supplier Thermo Scientific Chemicals Supplier No. 191650010

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51649-83-3
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.326
InChI Key YOAWSYSKQHLFPM-UHFFFAOYSA-N
Synonym 6-Aminofluorescein
PubChem CID 103924
IUPAC Name 5-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Beilstein 19, V,8, 713
Infrared Spectrum Authentic
Formula Weight 347.32
Percent Purity ≥96.0%
Color Brown to Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Fluoresceinamine isomer II, 97%
EINECSNumber 257-334-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.