Learn More
ProteinSimple Fluorescent 5x Master Mix 1
Supplier:  ProteinSimple PSFL018
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Pack includes 8 packets with one each of the Single Use FL Standard (29 kDa system control protein) and Single Use DTT vials. Compatible with 12-230 Separation Modules.
Catalog No. NC1189080